5-Methoxy-4-methylpyridine-3-boronic acid ≥95%

Supplier: AOBChem USA

Warning

23131-1G 23131-25G 23131-500MG 23131-5G
76938-232EA 439.22 USD
76938-232 76938-234 76938-236 76938-238
5-Methoxy-4-methylpyridine-3-boronic acid ≥95%
5-Methoxy-4-methylpyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD18258515
CAS Number: 1451391-97-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR