4-(4-Bromo-3-fluorobenzyl)morpholine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: N-(4-Bromo-3-fluorobenzyl)morpholine

23062-1G 23062-25G 23062-5G
76938-224EA 81.69 USD
76938-224 76938-226 76938-228
4-(4-Bromo-3-fluorobenzyl)morpholine ≥97%
4-(4-Bromo-3-fluorobenzyl)morpholine
Formula: C₁₁H₁₃BrFNO
MW: 274.133 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15526848
CAS Number: 897016-96-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR