Methyl 2,5-difluoro-4-hydroxybenzoate ≥97%

Supplier: AOBChem USA

Warning

23038-1G 23038-25G 23038-500MG 23038-5G
76938-200EA 184.6 USD
76938-200 76938-202 76938-204 76938-206
Methyl 2,5-difluoro-4-hydroxybenzoate ≥97%
Methyl 2,5-difluoro-4-hydroxybenzoate

Precaución: for Research Use Only

Formula: C₈H₆F₂O₃
MW: 188.128 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18399128
CAS Number: 1261684-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR