2-Morpholino-5-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(Morpholino)pyridine-5-boronic acid, 6-(Morpholino)pyridine-3-boronic acid, 6-Morpholinopyridine-3-boronic acid, 6-Morpholino-3-pyridineboronic Acid

22867-25G 22867-5G
76938-176EA 524.09 USD
76938-176 76938-178
2-Morpholino-5-pyridineboronic acid ≥97%
2-Morpholino-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₃BN₂O₃
MW: 208.0200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03086116
CAS Number: 904326-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR