6-Methoxy-4-methylpyridine-3-carboxylic acid ≥95%

Supplier: AOBChem USA

Warning

22680-1G 22680-25G 22680-500MG 22680-5G
76938-158EA 206.88 USD
76938-158 76938-160 76938-162 76938-164
6-Methoxy-4-methylpyridine-3-carboxylic acid ≥95%
6-Methoxy-4-methylpyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.162 g/mol
Storage Temperature: Ambient
CAS Number: 943719-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR