6-Bromo-2-fluoro-3-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

22605-1G 22605-25G 22605-5G
76938-144EA 62.6 USD
76938-144 76938-146 76938-148
6-Bromo-2-fluoro-3-methylbenzoic acid ≥97%
6-Bromo-2-fluoro-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.034 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20526384
CAS Number: 743466-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR