Potassium (3-fluoro-4-formylphenyl)trifluoroborate ≥97% research grade

Supplier: AOBChem USA

Warning

22540-25G 22540-5G
76938-120EA 595.17 USD
76938-120 76938-122
Potassium (3-fluoro-4-formylphenyl)trifluoroborate ≥97% research grade
Potassium (3-fluoro-4-formylphenyl)trifluoroborate
Formula: C₇H₄BF₄O·K
Storage Temperature: Ambient
MDL Number: MFCD09993334
CAS Number: 1451390-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR