4-Isoquinolinecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Isoquinoline-4-carboxylic acid

22315-25G 22315-5G
76938-094EA 381.93 USD
76938-094 76938-096
4-Isoquinolinecarboxylic acid ≥97%
4-Isoquinolinecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₇NO₂
MW: 173.1681 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00094344
CAS Number: 7159-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR