6-Bromo-2-fluoronicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-2-fluoro-3-pyridinecarboxylic acid

22125-1G 22125-25G 22125-5G
76938-066EA 99.73 USD
76938-066 76938-068 76938-070
6-Bromo-2-fluoronicotinic acid ≥97%
6-Bromo-2-fluoronicotinic acid
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185804
CAS Number: 1214345-17-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR