Methyl-5-chloro-2-methoxynicotinate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: Methyl 5-chloro-2-methoxynicotinate, Methyl 5-chloro-2-methoxy-3-pyridinecarboxylate

22118-25G 22118-5G
76938-054EA 431.79 USD
76938-054 76938-056
Methyl-5-chloro-2-methoxynicotinate ≥97% research grade
Methyl-5-chloro-2-methoxynicotinate
Formula: C₈H₈ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD12025883
CAS Number: 82060-51-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR