6-Chloro-5-methoxypyridine-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

21851-1G 21851-25G 21851-500MG 21851-5G
76938-024EA 292.81 USD
76938-024 76938-026 76938-028 76938-030
6-Chloro-5-methoxypyridine-3-boronic acid ≥97%
6-Chloro-5-methoxypyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD13189015
CAS Number: 1548827-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR