6-Bromo-2-fluoropyridine-3-carboxaldehyde ≥97%

Supplier: AOBChem USA

Warning

21817-1G 21817-25G 21817-500MG 21817-5G
76938-012EA 126.25 USD
76938-012 76938-014 76938-016 76938-018
6-Bromo-2-fluoropyridine-3-carboxaldehyde ≥97%
6-Bromo-2-fluoropyridine-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₃BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD16556268
CAS Number: 1227588-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR