5-Hydroxy-2-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

21756-1G 21756-25G 21756-500MG 21756-5G
76937-996EA 165.51 USD
76937-996 76937-998 76938-000 76938-002
5-Hydroxy-2-methylphenylboronic acid pinacol ester ≥97%
5-Hydroxy-2-methylphenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₃
Storage Temperature: Ambient
MDL Number: MFCD16994425
CAS Number: 1196985-65-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR