Methyl-3-fluoro-2-methoxybenzoate ≥97%

Supplier: AOBChem USA

Warning

21723-25G 21723-5G
76937-984EA 346.92 USD
76937-984 76937-986
Methyl-3-fluoro-2-methoxybenzoate ≥97%
Methyl-3-fluoro-2-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉FO₃
Storage Temperature: Ambient
MDL Number: MFCD09753133
CAS Number: 106428-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR