6-Butoxy-5-methylphenylpyridine-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

21670-1G 21670-25G 21670-5G
76937-972EA 63.66 USD
76937-972 76937-974 76937-976
6-Butoxy-5-methylphenylpyridine-3-boronic acid ≥97%
6-Butoxy-5-methylphenylpyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₆BNO₃
Storage Temperature: Ambient
MDL Number: MFCD18087689
CAS Number: 1256355-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR