6-Fluoro-3-hydroxy-2-methylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

21662-1G 21662-25G 21662-500MG 21662-5G
76937-958EA 147.47 USD
76937-958 76937-960 76937-962 76937-964
6-Fluoro-3-hydroxy-2-methylpyridine ≥97% research grade
6-Fluoro-3-hydroxy-2-methylpyridine
Formula: C₆H₆FNO
Storage Temperature: Refrigerator
MDL Number: MFCD16611088
CAS Number: 1227577-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR