Ethyl 6-chloro-5-methylpyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-5-methylnicotinic acid ethyl ester

21590-1G 21590-25G 21590-500MG 21590-5G
76937-944EA 155.96 USD
76937-944 76937-946 76937-948 76937-950
Ethyl 6-chloro-5-methylpyridine-3-carboxylate ≥97%
Ethyl 6-chloro-5-methylpyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₀ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD20485595
CAS Number: 942511-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR