5-Isopropoxy-2-methylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

21524-1G 21524-25G 21524-500MG 21524-5G
76937-900EA 173.99 USD
76937-900 76937-902 76937-904 76937-906
5-Isopropoxy-2-methylbenzoic acid ≥97%
5-Isopropoxy-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD19105378
CAS Number: 1266965-23-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR