Methyl-2,5-dichloroisonicotinate ≥95%

Supplier: AOBChem USA

Warning

21512-1G 21512-25G 21512-5G
76937-886EA 62.6 USD
76937-886 76937-888 76937-890
Methyl-2,5-dichloroisonicotinate ≥95%
Methyl 2,5-dichloroisonicotinate

Precaución: for Research Use Only

Formula: C₇H₅Cl₂NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09951944
CAS Number: 623585-74-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR