Methyl-5-methoxynicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 5-methoxypyridine-3-carboxylate, Methyl 5-methoxynicotinate

21450-25G 21450-5G
76937-880EA 584.56 USD
76937-880 76937-882
Methyl-5-methoxynicotinate ≥97%
Methyl-5-methoxynicotinate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
Storage Temperature: Ambient
MDL Number: MFCD09265109
CAS Number: 29681-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR