6-Chloro-2-Fluoro-3-Methanesulfonyl-Pyridine ≥97%

Supplier: AOBChem USA

Warning

21410-1G 21410-25G 21410-500MG 21410-5G
76937-868EA 173.99 USD
76937-868 76937-870 76937-872 76937-874
6-Chloro-2-Fluoro-3-Methanesulfonyl-Pyridine ≥97%
6-Chloro-2-Fluoro-3-Methanesulfonyl-Pyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFNO₂S
Storage Temperature: Refrigerator
MDL Number: MFCD16658094
CAS Number: 1093880-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR