6-Bromo-2-methoxypyridine-3-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-2-methoxypyridin-3-ylboronic acid

21375-1G 21375-25G 21375-5G
76937-854EA 62.6 USD
76937-854 76937-856 76937-858
6-Bromo-2-methoxypyridine-3-boronic acid ≥97%
6-Bromo-2-methoxypyridine-3-boronic acid
Formula: C₆H₇BBrNO₃
MW: 231.83 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18382838
CAS Number: 1242314-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR