Methyl 6-methoxy-2-methylpyridine-3-carboxylate ≥97%

Supplier: AOBChem USA

Warning

21300-1G 21300-25G 21300-500MG 21300-5G
76937-838EA 317.21 USD
76937-838 76937-840 76937-842 76937-844
Methyl 6-methoxy-2-methylpyridine-3-carboxylate ≥97%
Methyl 6-methoxy-2-methylpyridine-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.189 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16611640
CAS Number: 137347-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR