Methyl-6-methoxypicolinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Methoxy-pyridine-2-carboxylic acid methyl ester, Methyl 6-methoxypyridine-2-carboxylate, Methyl 6-methoxypicolinate

21270-25G 21270-5G
76937-814EA 310.85 USD
76937-814 76937-816
Methyl-6-methoxypicolinate ≥97%
Methyl 6-methoxypicolinate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07779517
CAS Number: 26256-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR