7-Methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

21016-1G 21016-25G 21016-5G
76937-786EA 70.02 USD
76937-786 76937-788 76937-790
7-Methyl-1H-indazole ≥97%
7-Methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.1600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05663434
CAS Number: 3176-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR