4-Iodo-3-(2,2,2-trimethylacetamido)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

20819-25G 20819-5G
76937-768EA 595.17 USD
76937-768 76937-770
4-Iodo-3-(2,2,2-trimethylacetamido)pyridine ≥97% research grade
N-(4-Iod-3-pyridyl)pivalamide
Formula: C₁₀H₁₃IN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD06659003
CAS Number: 113975-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR