6-Chloro-2-fluoronicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-2-fluoro-3-pyridinecarboxylic acid

20676-1G 20676-25G 20676-5G
76937-754EA 114.58 USD
76937-754 76937-756 76937-758
6-Chloro-2-fluoronicotinic acid ≥97%
6-Chloro-2-fluoronicotinic acid
Formula: C₆H₃ClFNO₂
MW: 175.543 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16610397
CAS Number: 1211578-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR