6-Chloro-2-fluoropyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

20673-1G 20673-25G 20673-5G
76937-740EA 62.6 USD
76937-740 76937-742 76937-744
6-Chloro-2-fluoropyridin-3-ol ≥97%
6-Chloro-2-fluoropyridin-3-ol

Precaución: for Research Use Only

Formula: C₅H₃ClFNO
Storage Temperature: Refrigerator
MDL Number: MFCD07437887
CAS Number: 883107-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR