Methyl 3,5-dibromoisonicotinate ≥95%

Supplier: AOBChem USA

Warning

Synonyms: Methyl 3,5-dibromo-4-pyridinecarboxylate

20628-1G 20628-25G 20628-500MG 20628-5G
76937-712EA 198.39 USD
76937-712 76937-714 76937-716 76937-718
Methyl 3,5-dibromoisonicotinate ≥95%
Methyl 3,5-dibromoisonicotinate
Formula: C₇H₅Br₂NO₂
MW: 294.92 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698026
CAS Number: 1214375-07-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR