Isoquinoline-5-carbaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
20573-25G
20573-5G
76937-704EA
403.15
USD
76937-704
76937-706
Isoquinoline-5-carbaldehyde ≥97%
Isoquinoline-5-carbaldehyde
Precaución: for Research Use Only
Formula:
C₁₀H₇NO Storage Temperature: Refrigerator |
MDL Number:
MFCD03412483 CAS Number: 80278-67-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...