Ethyl 2,5-dibromo-3-pyridinecarboxylate ≥97%

Supplier: AOBChem USA

Warning

20560-1G 20560-25G 20560-5G
76937-692EA 64.72 USD
76937-692 76937-694 76937-696
Ethyl 2,5-dibromo-3-pyridinecarboxylate ≥97%
Ethyl 2,5-dibromo-3-pyridinecarboxylate

Precaución: for Research Use Only

Formula: C₈H₇Br₂NO₂
MW: 308.955 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12032194
CAS Number: 1214375-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR