tert-Butyl 4-bromo-1-naphthoate ≥97%

Supplier: AOBChem USA

Warning

20482-1G 20482-25G 20482-500MG 20482-5G
76937-666EA 155.96 USD
76937-666 76937-668 76937-670 76937-672
tert-Butyl 4-bromo-1-naphthoate ≥97%
tert-Butyl 4-bromo-1-naphthoate

Precaución: for Research Use Only

Formula: C₁₅H₁₅BrO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09258636
CAS Number: 929000-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR