6-Methoxy-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

20399-25G 20399-5G
76937-656EA 293.87 USD
76937-656 76937-658
6-Methoxy-1H-indazole ≥97%
6-Methoxy-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD09261132
CAS Number: 3522-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR