6-Bromo-2-methylpyridin-3-ol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-3-hydoxy-2-methylpyridine

20396-1G 20396-25G 20396-5G
76937-642EA 84.88 USD
76937-642 76937-644 76937-646
6-Bromo-2-methylpyridin-3-ol ≥97%
6-Bromo-2-methylpyridin-3-ol

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.022 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11044277
CAS Number: 118399-86-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR