6-Bromo-2,3-difluorophenetole ≥97%

Supplier: AOBChem USA

Warning

19970-1G 19970-25G 19970-500MG 19970-5G
76937-566EA 168.69 USD
76937-566 76937-568 76937-570 76937-572
6-Bromo-2,3-difluorophenetole ≥97%
6-Bromo-2,3-difluorophenetole

Precaución: for Research Use Only

Formula: C₈H₇BrF₂O
MW: 237.041 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11656841
CAS Number: 1122410-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR