Methyl 2-bromo-6-chlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

19930-25G 19930-5G
76937-558EA 524.09 USD
76937-558 76937-560
Methyl 2-bromo-6-chlorobenzoate ≥97%
Methyl 2-bromo-6-chlorobenzoate
Formula: C₈H₆BrClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD03789160
CAS Number: 685892-23-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR