7-Chloro-1H-indazole-4-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

19883-1G 19883-25G 19883-500MG 19883-5G
76937-544EA 290.69 USD
76937-544 76937-546 76937-548 76937-550
7-Chloro-1H-indazole-4-boronic acid pinacol ester ≥97%
7-Chloro-1H-indazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BClN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD22628189
CAS Number: 1186334-62-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR