6-Bromo-2-fluoro-3-trifluoromethyl-benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Bromo-2-fluoro-3-(trifluoromethyl)benzaldehyde

19880-1G 19880-25G 19880-5G
76937-530EA 88.06 USD
76937-530 76937-532 76937-534
6-Bromo-2-fluoro-3-trifluoromethyl-benzaldehyde ≥97%
6-Bromo-2-fluoro-3-trifluoromethyl-benzaldehyde

Precaución: for Research Use Only

Formula: C₈H₃BrF₄O
MW: 271.006 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16658656
CAS Number: 1055176-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR