Methyl 5-bromo-2-cyanobenzoate ≥97%

Supplier: AOBChem USA

Warning

19701-1G 19701-25G 19701-500MG 19701-5G
76937-494EA 218.55 USD
76937-494 76937-496 76937-498 76937-500
Methyl 5-bromo-2-cyanobenzoate ≥97%
Methyl 5-bromo-2-cyanobenzoate
Formula: C₉H₆BrNO₂
MW: 240.05 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15523586
CAS Number: 714237-95-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR