5-Methoxy-2-methylbenzaldehyde ≥95%

Supplier: AOBChem USA

Warning

19575-1G 19575-25G 19575-5G
76937-484EA 71.09 USD
76937-484 76937-486 76937-488
5-Methoxy-2-methylbenzaldehyde ≥95%
5-Methoxy-2-methylbenzaldehyde
Formula: C₉H₁₀O₂
Storage Temperature: Refrigerator
MDL Number: MFCD12547802
CAS Number: 56724-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR