7-Bromo-1-methyl-1H-indazole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 7-Bromo-1-methylindazole

19298-1G 19298-25G 19298-5G
76937-462EA 66.84 USD
76937-462 76937-464 76937-466
7-Bromo-1-methyl-1H-indazole ≥97%
7-Bromo-1-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
Storage Temperature: Refrigerator
MDL Number: MFCD09878570
CAS Number: 1000576-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR