Ethyl-6-bromonicotinate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: Ethyl 2-bromo-5-pyridinecarboxylate, Ethyl 6-bromonicotinate, 6-Bromonicotinic acid ethyl ester

19274-25G 19274-5G
76937-454EA 254.62 USD
76937-454 76937-456
Ethyl-6-bromonicotinate ≥97%
Ethyl-6-bromonicotinate
Formula: C₈H₈BrNO₂
MW: 230.0586 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111853
CAS Number: 132334-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR