6-Methoxy-2-methylpyridine-3-boronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Methoxy-2-methyl-pyridine-3-boronic acid pinacol ester

19141-1G 19141-25G 19141-500MG 19141-5G
76937-430EA 233.4 USD
76937-430 76937-432 76937-434 76937-436
6-Methoxy-2-methylpyridine-3-boronic acid pinacol ester ≥97%
6-Methoxy-2-methylpyridine-3-boronic acid pinacol ester
Formula: C₁₃H₂₀BNO₃
MW: 249.11 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12923398
CAS Number: 1080028-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR