2-(4-Bromobenzyl)isoindoline-1,3-dione ≥97%

Supplier: AOBChem USA

Warning

Synonyms: N-(4-Bromobenzyl)phthalimide

19060-1G 19060-25G 19060-5G
76937-416EA 114.58 USD
76937-416 76937-418 76937-420
2-(4-Bromobenzyl)isoindoline-1,3-dione ≥97%
2-(4-Bromobenzyl)isoindoline-1,3-dione
Formula: C₁₅H₁₀BrNO₂
MW: 316.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04210206
CAS Number: 153171-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR