5-Methyl-2-(2,2,2-trimethylacetamido)pyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: N-(5-Methylpyridin-2-yl)pivalamide

18853-1G 18853-25G 18853-5G
76937-398EA 109.28 USD
76937-398 76937-400 76937-402
5-Methyl-2-(2,2,2-trimethylacetamido)pyridine ≥97%
5-Methyl-2-(2,2,2-trimethylacetamido)pyridine
Formula: C₁₁H₁₆N₂O
MW: 192.262 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03374646
CAS Number: 86847-78-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR