tert-Butyl 4-bromo-3-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
18715-1G
18715-25G
18715-500MG
18715-5G
76937-360EA
120.95
USD
76937-360
76937-362
76937-364
76937-366
tert-Butyl 4-bromo-3-methylbenzoate ≥97%
tert-Butyl 4-bromo-3-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₂H₁₅BrO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD08703429 CAS Number: 347174-28-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...