tert-Butyl-4-bromo-2-chlorobenzoate ≥97%

Supplier: AOBChem USA

Warning

Synonyms: tert-Butyl 4-bromo-2-chlorobenzoate

18713-1G 18713-25G 18713-5G
76937-346EA 62.6 USD
76937-346 76937-348 76937-350
tert-Butyl-4-bromo-2-chlorobenzoate ≥97%
tert-Butyl 4-bromo-2-chlorobenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₂BrClO₂
Storage Temperature: Refrigerator
MDL Number: MFCD08703428
CAS Number: 929000-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR