6-Chloro-2-methylnicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-2-methylpyridine-3-carboxylic acid

18640-25G 18640-5G
76937-338EA 201.58 USD
76937-338 76937-340
6-Chloro-2-methylnicotinic acid ≥97%
6-Chloro-2-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD09842452
CAS Number: 137129-98-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR