Benzenamine, 2-bromo-N-(diphenylmethylene)- ≥95%

Supplier: AOBChem USA

Warning

18281-1G 18281-25G 18281-5G
76937-292EA 62.6 USD
76937-292 76937-294 76937-296
Benzenamine, 2-bromo-N-(diphenylmethylene)- ≥95%
Benzenamine, 2-bromo-N-(diphenylmethylene)-

Precaución: for Research Use Only

Formula: C₁₉H₁₄BrN
Storage Temperature: Ambient
MDL Number: MFCD31619395
CAS Number: 251320-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR