N-(4-Methylphenyl)-2,2-dimethylpropanamide ≥97%

Supplier: AOBChem USA

Warning

18242-1G 18242-25G 18242-500MG 18242-5G
76937-282EA 213.25 USD
76937-282 76937-284 76937-286 76937-288
N-(4-Methylphenyl)-2,2-dimethylpropanamide ≥97%
N-(4-Methylphenyl)-2,2-dimethylpropanamide

Precaución: for Research Use Only

Formula: C₁₂H₁₇NO
Storage Temperature: Ambient
CAS Number: 21354-40-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR